Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598342 Supplier Invitrogen™ Supplier No. PA598342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDR1 Polyclonal Antibody for Western Blot, ELISA

NDR1 is involved in disease resistance. It is required for resistance conferred by multiple R genes recognizing different bacterial and oomycete pathogen isolates like avirulent P.syringae or H.parasitica (downy mildew). NDR1 is also required for the establishment of hypersensitive response (HR) and systemic acquired resistance (SAR) after infection with the bacterial pathogen P.syringae DC3000 carrying avrRpt2 and is required for resistance to the soilborne fungus V.longisporum. Interaction with RIN4 is required for the activation of the R gene RPS2 and RPS2-mediated resistance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDR1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NDR1
Gene Alias At3g20600; AtNDR1; CHUNP6892; K10D20.24; Late embryogenesis abundant (LEA) hydroxyproline-rich glycoprotein family; member of the nodal subgroup of the TGF-beta superfamily of peptide growth factors; NDR1; Ndr-1; NDR2; nodal-related 1; nodal-related-2; non race-specific disease resistance 1; NON-RACE SPECIFIC DISEASE RESISTANCE PROTEIN; non-race specific disease resistance protein (NDR1); Non-race specific disease resistance protein 1; nr-2; Protein NDR1; sqt; squint; squint precursor; Znr-1 related; znr2; Znr-2
Gene Symbols NDR1
Host Species Rabbit
Immunogen Recombinant Zebrafish Nodal-related 1 protein (71-213AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 799352
Target Species Zebrafish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.