Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Mouse anti-Human, Clone: 11G4, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$340.50 - $499.50

Specifications

Antigen NDRG1
Clone 11G4
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1025, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25941550
SDP
View Documents
Novus Biologicals
NBP25941550UL
50 μL
Each for $340.50
Only null left
Add to Cart
 
NBP259415X
SDP
View Documents
Novus Biologicals
NBP259415100UL
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

NDRG1 Monoclonal specifically detects NDRG1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NDRG1
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Cancer
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
10397
Recombinant human NDRG1 (1-394aa) purified from E. coli
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
11G4
Western Blot 1:1000, Flow Cytometry 1:10-1:1025, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Monoclonal
Purified
RUO
Human
CAP43, CMT4D, Differentiation-related gene 1 protein, DRG-1, DRG1HMSNL, NDR1, Nickel-specific induction protein Cap43, NMSL, N-myc downstream regulated 1, N-myc downstream-regulated gene 1 protein, protein NDRG1, protein regulated by oxygen-1, PROXY1, Reducing agents and tunicamycin-responsive protein, RIT42, RTPGC4, TARG1, TDD5, tunicamycin-responsive protein
NDRG1
IgG1 κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.