Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDRG1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 269021AP150UL

Description
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein involved in stress responses, hormone responses, cell growth, and differentiation. It is necessary for p53-mediated caspase activation and apoptosis. Mutation in this gene has been reported to be causative for hereditary motor and sensory neuropathy-Lom. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
NDRG1 | |
Polyclonal | |
Unconjugated | |
NDRG1 | |
CAP43, CMT4D, DRG 1, DRG1, GC4, HMSNL, N myc downstream regulated 1, NDR1, NDRG1, Protein NDRG1, PROXY1, RIT42, RTP, TARG1, TDD5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10397 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92597 | |
NDRG1 | |
NDRG1 Fusion Protein Ag25424 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction