Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDRG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155431
Description
NDRG2 Polyclonal antibody specifically detects NDRG2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
NDRG2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
NDRG2 | |
Synthetic peptides corresponding to NDRG2(NDRG family member 2) The peptide sequence was selected from the C terminal of NDRG2. Peptide sequence GYMASSCMTRLSRSRTASLTSAASVDGNRSRSRTLSQSSESGTLSSGPPG. | |
Affinity purified | |
RUO | |
Primary | |
Zebrafish: 80%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
DKFZp781G1938, FLJ25522, KIAA1248cytoplasmic protein Ndr1, NDR1-related protein NDR2, NDRG family member 2, protein NDRG2, Protein Syld709613, syld709613 protein, SYLDN-myc downstream regulator 2 | |
Rabbit | |
35 kDa | |
100 μL | |
Cancer | |
57447 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction