Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDRG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102726 Supplier Invitrogen™ Supplier No. PA5102726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NDRG2.

NDRG2's gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. NDRG2 is a cytoplasmic protein that may play a role in neurite outgrowth. Its gene may be involved in glioblastoma carcinogenesis.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDRG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NDRG2
Gene Accession No. Q8VBU2, Q9QYG0, Q9UN36
Gene Alias AI182517; antidepressant-related protein ADRG123; AU040374; cytoplasmic protein Ndr1; DKFZp781G1938; FLJ25522; Kiaa1248; NDR1-related protein NDR2; Ndr2; NDRG family member 2; NDRG1-related protein; NDRG2; NDRG2var; N-myc downstream regulated 2; N-Myc downstream regulated gene 2; N-myc downstream regulator 2; N-myc downstream-regulated gene 2; N-myc downstream-regulated gene 2 protein; Protein Ndr2; protein NDRG2; Protein Syld709613; SYLD; syld709613 protein
Gene Symbols NDRG2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NDRG2(Accession Q9UN36), corresponding to amino acid residues R327-C371.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171114, 29811, 57447
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.