Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ NDRG2 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142707
                        
                            
                        
                        
                        
                            
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
Additional Information: This antibody is expected to recognize reported isoform a (NP_963293.1) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
NDRG2's gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. NDRG2 is a cytoplasmic protein that may play a role in neurite outgrowth. Its gene may be involved in glioblastoma carcinogenesis.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
            Specifications
| NDRG2 | |
| Polyclonal | |
| Unconjugated | |
| NDRG2 | |
| AI182517; antidepressant-related protein ADRG123; AU040374; cytoplasmic protein Ndr1; DKFZp781G1938; FLJ25522; Kiaa1248; NDR1-related protein NDR2; Ndr2; NDRG family member 2; NDRG1-related protein; NDRG2; NDRG2var; N-myc downstream regulated 2; N-Myc downstream regulated gene 2; N-myc downstream regulator 2; N-myc downstream-regulated gene 2; N-myc downstream-regulated gene 2 protein; Protein Ndr2; protein NDRG2; Protein Syld709613; SYLD; syld709613 protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 171114, 29811, 57447 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8VBU2, Q9QYG0, Q9UN36 | |
| NDRG2 | |
| Peptide with sequence KEAELAARILLDQGQ-C (aa 25-39). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction