Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | NDRG2 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7643
![]() ![]() |
Proteintech Group Inc
120151AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
NDRG2′s gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. NDRG2 is a cytoplasmic protein that may play a role in neurite outgrowth. Its gene may be involved in glioblastoma carcinogenesis.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
NDRG2 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8VBU2, Q9QYG0, Q9UN36 | |
171114, 29811, 57447 | |
NDRG2 Fusion Protein Ag2629 | |
Primary | |
-20°C | |
NDRG2 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1248, NDRG family member 2, NDRG2, Protein NDRG2, Protein Syld709613, SYLD | |
Ndrg2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title