Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDRG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553766 Supplier Invitrogen™ Supplier No. PA553766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CAKGWIDWAA SKLSGLTTNV VDIILAHHFG QEELQANLDL IQTYRMHIAQ DINQDNLQLF LNSYNGRRDL EIERPILGQN DNKSKTLKCS TLLVVGD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 87%.

NDRG3 (NDRG Family Member 3) is a Protein Coding gene. Diseases associated with NDRG3 includeCharcot-Marie-Tooth Disease, Type 4DandNeuropathy, Congenital Hypomyelinating Or Amyelinating, Autosomal Recessive. Among its related pathways areApoptosis and AutophagyandPI3K / Akt Signaling. An important paralog of this gene isNDRG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDRG3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDRG3
Gene Accession No. Q9UGV2
Gene Alias 4833415O14Rik; AI747193; Ndr3; NDRG family member 3; Ndrg3; N-myc downstream regulated 3; N-myc downstream regulated gene 3; N-myc downstream regulated gene 3-like; N-myc downstream-regulated gene 3 protein; Protein Ndr3; protein NDRG3
Gene Symbols NDRG3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDRG3. Recombinant protein control fragment (Product #RP-92282).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57446
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.