Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDRG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579723
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579723 Supplier Invitrogen™ Supplier No. PA579723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: COLO320 whole cell. IHC: rat brain tissue, human mammary cancer tissue, human intestinal cancer tissue, mouse brain tissue.

NDRG3 (NDRG Family Member 3) is a Protein Coding gene. Diseases associated with NDRG3 includeCharcot-Marie-Tooth Disease, Type 4DandNeuropathy, Congenital Hypomyelinating Or Amyelinating, Autosomal Recessive. Among its related pathways areApoptosis and AutophagyandPI3K / Akt Signaling. An important paralog of this gene isNDRG1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDRG3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene NDRG3
Gene Accession No. Q6AYR2, Q9QYF9, Q9UGV2
Gene Alias 4833415O14Rik; AI747193; Ndr3; NDRG family member 3; Ndrg3; N-myc downstream regulated 3; N-myc downstream regulated gene 3; N-myc downstream regulated gene 3-like; N-myc downstream-regulated gene 3 protein; Protein Ndr3; protein NDRG3
Gene Symbols NDRG3
Host Species Rabbit
Immunogen E.coli-derived human NDRG3 recombinant protein (Position: L181-R239).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296318, 29812, 57446
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.