Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDRG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550095
Description
The antibody detects endogenous levels of total NDRG4 protein.
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signaling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
NDRG4 | |
Polyclonal | |
Unconjugated | |
NDRG4 | |
BDM1; Brain development-related molecule 1; D8Bwg1337e; development-related protein; DKFZp686I1615; FLJ30586; FLJ42011; KIAA1180; MGC19632; Ndr1-rs; Ndr4; NDRG family member 4; NDRG4; N-myc downstream regulated 1, related sequence; N-myc downstream regulated 4; N-myc downstream regulated gene 4; N-myc downstream-regulated gene 4 protein; Protein Ndr4; protein NDRG4; R74996; smap8; SMAP-8; smooth muscle-associated protein 8; vascular smooth muscle cell-associated protein 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
234593, 64457, 65009 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BTG7, Q9ULP0, Q9Z2L9 | |
NDRG4 | |
Synthesized peptide derived from internal of human NDRG4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction