Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4396 Supplier Bioss Supplier No. BS1999R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signaling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.

Specifications

Antigen NDRG4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NDRG4
Gene Accession No. Q9ULP0
Gene Alias BDM1; Brain development-related molecule 1; D8Bwg1337e; development-related protein; DKFZp686I1615; FLJ30586; FLJ42011; KIAA1180; MGC19632; Ndr1-rs; Ndr4; NDRG family member 4; Ndrg4; N-myc downstream regulated 1, related sequence; N-myc downstream regulated 4; N-myc downstream regulated gene 4; N-myc downstream-regulated gene 4 protein; Protein Ndr4; Protein NDRG4; R74996; smap8; SMAP-8; smooth muscle-associated protein 8; Vascular smooth muscle cell-associated protein 8
Gene Symbols NDRG4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NDRG4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65009
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.