Learn More
Invitrogen™ NDST1 Polyclonal Antibody

Description
Positive controls that may be used include: U937 Cell Lysate.
Specifications
Specifications
| Antigen | NDST1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NDST1 |
| Gene Accession No. | P52848 |
| Gene Alias | [Heparan sulfate]-glucosamine N-sulfotransferase 1; 1200015G06Rik; b2b2230Clo; bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 1; Glucosaminyl N-deacetylase/N-sulfotransferase 1; Heparan sulfate N-deacetylase 1; Heparan sulfate N-sulfotransferase 1; heparan sulfate/heparin GlcNAc N-deacetylase/GlcN N-sulfotransferase 1; heparan sulfate-N-deacetylase/N-sulfotransferase; HSNST 1; HSST; HSST1; MRT46; N-deacetylase and N-sulfotransferase 1; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1; N-deacetylase/N-sulfotransferase 1; N-deacetylase/N-sulfotransferase-1; N-Deacetylase-N-sulfotransferase 1; Ndst1; NDST-1; N-heparan sulfate sulfotransferase 1; N-HSST 1; NST1 |
| Gene Symbols | NDST1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.