Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDST1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9891R

Description
Essential bifunctional enzyme that catalyzes both the N- deacetylation and the N-sulfation of glucosamine (GlcNAc) of the glycosaminoglycan in heparan sulfate. Modifies the GlcNAc-GlcA disaccharide repeating sugar backbone to make N-sulfated heparosan, a prerequisite substrate for later modifications in heparin biosynthesis. Plays a role in determining the extent and pattern of sulfation of heparan sulfate. Compared to other NDST enzymes, its presence is absolutely required. Participates in biosynthesis of heparan sulfate that can ultimately serve as L- selectin ligands, thereby playing a role in inflammatory response.Specifications
NDST1 | |
Polyclonal | |
Unconjugated | |
NDST1 | |
[Heparan sulfate]-glucosamine N-sulfotransferase 1; 1200015G06Rik; b2b2230Clo; Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 1; Glucosaminyl N-deacetylase/N-sulfotransferase 1; Heparan sulfate N-deacetylase 1; Heparan sulfate N-sulfotransferase 1; heparan sulfate/heparin GlcNAc N-deacetylase/GlcN N-sulfotransferase 1; heparan sulfate-N-deacetylase/N-sulfotransferase; HSNST 1; HSST; HSST1; MRT46; N-deacetylase and N-sulfotransferase 1; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1; N-deacetylase/N-sulfotransferase 1; N-deacetylase/N-sulfotransferase-1; N-Deacetylase-N-sulfotransferase 1; Ndst1; NDST-1; N-heparan sulfate sulfotransferase 1; N-HSST 1; NST1 | |
Rabbit | |
Protein A | |
RUO | |
3340 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P52848 | |
NDST1 | |
KLH conjugated synthetic peptide derived from human NDST1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction