Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDST2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572813
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572813 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572813 Supplier Invitrogen™ Supplier No. PA572813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDST2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

NDST2 is a member of theN-deacetylase/N-sulfotransferase subfamily of the sulfotransferase1 proteins. The encoded enzyme has dual functions in processing glucosamine and heparin polymers, including N-deacetylation andN-sulfation. The encoded protein may be localized to the Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDST2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NDST2
Gene Accession No. P52849, P52850
Gene Alias [Heparan sulfate]-glucosamine N-sulfotransferase; bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 2; Glucosaminyl N-deacetylase/N-sulfotransferase 2; glucosaminyl N-deacetylase/N-sulphotransferase 2; heparan glucosaminyl N-deacetylase/N-sulfotransferase 2; heparan N-sulfotransferase 2; Heparan sulfate N-deacetylase 2; heparan sulfate N-deacetylase/N-sulfotransferase 2; Heparan sulfate N-sulfotransferase 2; HSST2; mastocytoma N-deacetylase/N-sulfotransferase; Mndns; N-deacetylase and N-sulfotransferase 2; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2; N-deacetylase/N-sulfotransferase 2; Ndst2; NDST-2; N-heparan sulfate sulfotransferase 2; N-HSST 2; NST2
Gene Symbols NDST2
Host Species Rabbit
Immunogen NDST2 antibody for a 16 amino acid peptide near the amino terminus of human NDST2. The immunogen is within amino acids 20 - 70 of NDST2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114002, 17423, 8509
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.