Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDST3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528057
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (92%), Rat (91%), Rhesus Monkey (99%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the heparan sulfate/heparin GlcNAc N-deacetylase/ N-sulfotransferase family. The encoded enzyme is a type II transmembrane protein that resides in the Golgi apparatus. This monomeric bifunctional enzyme catalyzes the N-deacetylation and N-sulfation of N-acetylglucosamine residues in heparan sulfate and heparin, which are the initial chemical modifications required for the biosynthesis of the functional oligosaccharide sequences that define the specific ligand binding activities of heparan sulfate and heparin.
Specifications
| NDST3 | |
| Polyclonal | |
| Unconjugated | |
| NDST3 | |
| Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 3; GlcNAc N-deacetylase/ N-sulfotransferase 3; glucosaminyl N-deacetylase/N-sulfotransferase 3; Heparan sulfate N-deacetylase 3; Heparan sulfate N-sulfotransferase 3; hNDST-3; HSST3; N-deacetylase and N-sulfotransferase 3; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3; N-deacetylase/N-sulfotransferase 3; NDST3; NDST-3; N-heparan sulfate sulfotransferase 3; N-HSST 3; UNQ2544/PRO4998 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95803 | |
| NDST3 | |
| Recombinant protein encompassing a sequence within the center region of human NDST3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction