Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDST4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552794 Supplier Invitrogen™ Supplier No. PA552794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYKQEMTLIE TTAEAECTDI KILPYRSMEL KTVKPIDTSK TDPTVLLFVE SQYSQLGQDI IAILESSRFQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

Essential bifunctional enzyme that catalyzes both the N-deacetylation and the N-sulfation of glucosamine (GlcNAc) of the glycosaminoglycan in heparan sulfate. Modifies the GlcNAc-GlcA disaccharide repeating sugar backbone to make N-sulfated heparosan, a prerequisite substrate for later modifications in heparin biosynthesis. Has low deacetylase activity but high sulfotransferase activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDST4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDST4
Gene Accession No. Q9H3R1
Gene Alias Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 4; glucosaminyl N-deacetylase/N-sulfotransferase 4; Heparan sulfate N-deacetylase 4; Heparan sulfate N-sulfotransferase 4; HSST4; N-deacetylase and N-sulfotransferase 4; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 4; N-deacetylase/N-sulfotransferase 4; NDST4; NDST-4; N-heparan sulfate sulfotransferase 4; N-HSST; N-HSST 4; NHSST4
Gene Symbols NDST4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDST4. Recombinant protein control fragment (Product #RP-90268).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.