Learn More
Invitrogen™ NDUFA1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total NDUFA1.
Specifications
Specifications
| Antigen | NDUFA1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | NDUFA1 |
| Gene Accession No. | O15239, O35683 |
| Gene Alias | 1810049F12Rik; CI-MWFE; complex I MWFE subunit; complex I-MWFE; MWFE; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1 (7.5kD, MWFE); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1, 7.5kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 1; NADH dehydrogenase 1 alpha subcomplex, 1; NADH oxidoreductase subunit MWFE; NADH ubiquinone oxidoreductase subunit MWFE; NADH:ubiquinone oxidoreductase (complex 1); NADH:ubiquinone oxidoreductase subunit A1; NADH-ubiquinone oxidoreductase MWFE subunit; NDUFA1; RGD1560955; type I dehydrogenase; ZNF183 |
| Gene Symbols | NDUFA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.