Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597007 Supplier Invitrogen™ Supplier No. PA597007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MWFEILPGLS VMGVCLLIPG LATAYIHRFT NGGKEKRVAH FGYHWSLMER DRRISGVDRY YVSKGLENID; Positive Samples: MCF-7, Mouse kidney, Mouse heart, Rat brain; Cellular Location: Matrix side, Mitochondrion inner membrane, Single-pass membrane protein.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFA1
Gene Accession No. O15239, O35683
Gene Alias 1810049F12Rik; CI-MWFE; complex I MWFE subunit; complex I-MWFE; MWFE; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1 (7.5kD, MWFE); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1, 7.5kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 1; NADH dehydrogenase 1 alpha subcomplex, 1; NADH oxidoreductase subunit MWFE; NADH ubiquinone oxidoreductase subunit MWFE; NADH:ubiquinone oxidoreductase (complex 1); NADH:ubiquinone oxidoreductase subunit A1; NADH-ubiquinone oxidoreductase MWFE subunit; NDUFA1; RGD1560955; type I dehydrogenase; ZNF183
Gene Symbols NDUFA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human NDUFA1 (NP_0045321).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363441, 4694, 54405
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.