Learn More
Invitrogen™ NDUFA10 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse kidney. Predicted reactivity: Chimpanzee (99%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | NDUFA10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.82 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | NDUFA10 |
| Gene Accession No. | O95299, Q99LC3 |
| Gene Alias | 2900053E13Rik; CI-42k; CI-42KD; complex I 42kDa subunit; complex I-42kD; FLJ21923; MGC5103; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial; NADH:ubiquinone oxidoreductase subunit A10; NADH-ubiquinone oxidoreductase 42 kDa subunit; Ndufa10; ndufa10l |
| Gene Symbols | NDUFA10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.