Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ndufa10l1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569770 Supplier Invitrogen™ Supplier No. PA569770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 83%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 79%.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ndufa10l1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ndufa10l1
Gene Accession No. 0
Gene Alias CI-42kD; Complex I-42kD; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10-like; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10-like 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial; NADH dehydrogenase 1 alpha subcomplex 10-like protein; NADH-ubiquinone oxidoreductase 42 kDa subunit; Ndufa10; Ndufa10l1
Gene Symbols Ndufa10l1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat Ndufa10l1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316632
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.