Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567228 Supplier Invitrogen™ Supplier No. PA567228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLNPPGTFLEG VAKVGQYTFT Highest antigen sequence identity to the following orthologs - mouse 62%, rat 57%.

NDUFA11 encodes a subunit of the membrane-bound mitochondrial complex I. Complex I is composed of numerous subunits and functions as the NADH-ubiquinol reductase of the mitochondrial electron transport chain. Mutations in this gene are associated with severe mitochondrial complex I deficiency. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFA11
Gene Accession No. Q86Y39
Gene Alias 2010012C24Rik; AV006275; B14.7; CI-B14.7; complex I B14.7 subunit; complex I-B14.7; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 11, 14.7kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 11; NADH:ubiquinone oxidoreductase subunit A11; NADH-ubiquinone oxidoreductase subunit B14.7; NDUFA11
Gene Symbols NDUFA11
Host Species Rabbit
Immunogen Recombinant Human NDUFA11. Recombinant protein control fragment (Product #RP-105555).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.