Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110018 Supplier Invitrogen™ Supplier No. PA5110018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAPKVFRQYW DIPDGTDCHR KAYSTTSIAS VAGLTAAAYR VTLNPPGTFL EGVAKVGQYT FTAAAVGAVF GLTTCISAHV REKPDDPLNY FLGGCAGGLT.

NDUFA11 encodes a subunit of the membrane-bound mitochondrial complex I. Complex I is composed of numerous subunits and functions as the NADH-ubiquinol reductase of the mitochondrial electron transport chain. Mutations in this gene are associated with severe mitochondrial complex I deficiency. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NDUFA11
Gene Accession No. Q9D8B4
Gene Alias 2010012C24Rik; AV006275; B14.7; CI-B14.7; complex I B14.7 subunit; complex I-B14.7; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 11, 14.7kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 11; NADH:ubiquinone oxidoreductase subunit A11; NADH-ubiquinone oxidoreductase subunit B14.7; NDUFA11
Gene Symbols NDUFA11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NDUFA11 (NP_783313.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69875
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.