Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | NDUFA12 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NDUFA12 Polyclonal specifically detects NDUFA12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NDUFA12 | |
Polyclonal | |
Rabbit | |
Human | |
13 kDa differentiation-associated protein, B17.2,13kDa differentiation-associated protein, CIB17.2, CI-B17.2, complex I B17.2 subunit, Complex I-B17.2, DAP13NADH-ubiquinone oxidoreductase subunit B17.2, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12, NADH: ubiquinone oxidoreductase | |
NDUFA12 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
55967 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MELVQVLKRGLQQITGHGGLRGYLRVFFRTNDAKVGTLVGEDKYGNKYYEDNKQFFGRHRWVVYTTEMN | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title