Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546212 Supplier Invitrogen™ Supplier No. PA546212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VLKRGLQQIT GHGGLRGYLR VFFRTNDAKV GTLVGEDKYG NKYYEDNKQF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Sheep: 100%; Yeast: 91%; Zebrafish: 100%.

This gene encodes a protein which is part of mitochondrial complex 1, part of the oxidative phosphorylation system in mitochondria. Complex 1 transfers electrons to ubiquinone from NADH which establishes a proton gradient for the generation of ATP. Mutations in this gene are associated with Leigh syndrome due to mitochondrial complex 1 deficiency. Pseudogenes of this gene are located on chromosomes 5 and 13. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ndufa12
Gene Accession No. Q9UI09
Gene Alias 13 kDa differentiation-associated protein; 13kDa differentiation-associated protein; 2410011G03Rik; AW112974; B17.2; CIB17.2; CI-B17.2; complex I B17.2 subunit; complex I-B17.2; DAP13; DAP13 protein; hypothetical protein LOC613140; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12; NADH:ubiquinone oxidoreductase b17.2 subunit; NADH:ubiquinone oxidoreductase subunit A12; NADH-ubiquinone oxidoreductase subunit B17.2; Ndufa12; RGD1311462; si:dkey-183c16.3; zgc:112053
Gene Symbols Ndufa12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NDUFA12 (aa 6-55).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55967
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.