Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596981 Supplier Invitrogen™ Supplier No. PA596981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MELVQVLKRG LQQITGHGGL RGYLRVFFRT NDAKVGTLVG EDKYGNKYYE DNKQFFGRHR WVVYTTEMNG KNTFWDVDGS MVPPEWHRWL HSMTDDPPTT KPLTARKFIW TNHKFNVTGT PEQYVPYSTT RKKIQEWIPP STPYK; Positive Samples: 293T, PC-12, HepG2, Mouse kidney, Mouse heart, Mouse testis, Rat heart, Rat brain; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.

This gene encodes a protein which is part of mitochondrial complex 1, part of the oxidative phosphorylation system in mitochondria. Complex 1 transfers electrons to ubiquinone from NADH which establishes a proton gradient for the generation of ATP. Mutations in this gene are associated with Leigh syndrome due to mitochondrial complex 1 deficiency. Pseudogenes of this gene are located on chromosomes 5 and 13. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ndufa12
Gene Accession No. Q7TMF3, Q9UI09
Gene Alias 13 kDa differentiation-associated protein; 13kDa differentiation-associated protein; 2410011G03Rik; AW112974; B17.2; CIB17.2; CI-B17.2; complex I B17.2 subunit; complex I-B17.2; DAP13; DAP13 protein; hypothetical protein LOC613140; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12; NADH:ubiquinone oxidoreductase b17.2 subunit; NADH:ubiquinone oxidoreductase subunit A12; NADH-ubiquinone oxidoreductase subunit B17.2; Ndufa12; RGD1311462; si:dkey-183c16.3; zgc:112053
Gene Symbols Ndufa12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human NDUFA12 (NP_0613261).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299739, 55967, 66414
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.