Learn More
Invitrogen™ NDUFA13 Monoclonal Antibody (6E1BH7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 438900
Description
Positive control: Human heart, Bovine heart, Rat heart, and Mouse heart isolated mitochondria, Human fibroblasts, HeLa cells.
A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.
Specifications
| NDUFA13 | |
| Monoclonal | |
| 1 mg/mL | |
| HEPES buffered saline with 0.02% sodium azide | |
| Q95KV7, Q9ERS2, Q9P0J0 | |
| Ndufa13, Ndufa13-ps1 | |
| Full-length recombinant human GRIM-19. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 6E1BH7 | |
| Unconjugated | |
| Ndufa13 | |
| 2700054G14Rik; AU022060; B16.6; CDA016; Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CGI-39; CI-B16.6; complex I B16.6 subunit; complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; gene associated with retinoic and interferon-induced mortality 19 protein; Gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM19; GRIM-19; LOC100911483; mitochondrial NADH:ubiquinone oxidoreductase B16.6 subunit; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13-like; NADH:ubiquinone oxidoreductase subunit A13; NADH-ubiquinone oxidoreductase B16.6 subunit; Ndufa13; YjeF N-terminal domain-containing protein 3; YjeF_N3; Yjefn3 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100911483, 314759, 338084, 51079, 67184 | |
| 4°C, do not freeze | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.