Learn More
Invitrogen™ NDUFA2 Monoclonal Antibody (6E7)

Description
Peptide Sequence: MAAAAASRGV GAKLGLREIR IHLCQRSPGS QGVRDFIEKR YVELKKANPD LPILIRECSD VQPKLWARYA FGQETNVPLN NFSADQVTRA LENVLSGKA.
Specifications
Specifications
| Antigen | NDUFA2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 6E7 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NDUFA2 |
| Gene Accession No. | O43678 |
| Gene Alias | AV000592; B8; C1-B8; CCDC76; CD14; CIB8; CI-B8; complex I B8 subunit; complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH:ubiquinone oxidoreductase subunit A2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8; NDUFA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.