Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA2 Monoclonal Antibody (OTI5A5)

NDUFA2 Monoclonal Antibody (OTI5A5)

Supplier:  Thermo Scientific MA527474

Catalog No. PIMA527474


Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NDUFA2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O43678
NDUFA2
Human recombinant protein fragment corresponding to amino acids 2-99 of NDUFA2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin)
OTI5A5
Unconjugated
NDUFA2
AV000592; B8; C1-B8; CCDC76; CD14; CIB8; CI-B8; complex I B8 subunit; complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH:ubiquinone oxidoreductase subunit A2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8; NDUFA2
Mouse
Affinity Chromatography
RUO
4695
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.