Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557469 Supplier Invitrogen™ Supplier No. PA557469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGVGAKLGLR EIRIHLCQRS PGSQGVRDFI EKRYVELKKA NPDLPILIRE CSDVQPKLWA RYAFGQETNV PLNNFSADQV TRALENVLSG K Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

The NDUFA2 gene encodes one of the accessory subunits of complex I, the first and largest complex of the mitochondrial respiratory chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFA2
Gene Accession No. O43678
Gene Alias AV000592; B8; C1-B8; CCDC76; CD14; CIB8; CI-B8; complex I B8 subunit; complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH:ubiquinone oxidoreductase subunit A2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8; NDUFA2
Gene Symbols NDUFA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFA2. Recombinant protein control fragment (Product #RP-96844).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4695
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.