Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550068 Supplier Invitrogen™ Supplier No. PA550068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total NDUFA4 protein.

Cytochrome c oxidase (COX, complex IV) is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. Required for complex IV maintenance. {ECO:0000269; PubMed:22902835}.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NDUFA4
Gene Accession No. O00483, Q62425
Gene Alias CI-9k; CI-MLRQ; Complex I 9kDa subunit; complex I-MLRQ; Cytochrome c oxidase subunit NDUFA4; EC 1.6.5.3; EC 1.6.99.3; MLRQ; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4 (9 kDa, MLRQ); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4; NADH-ubiquinone oxidoreductase MLRQ subunit; NDUFA4; NDUFA4, mitochondrial complex associated
Gene Symbols NDUFA4
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human NDUFA4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17992, 4697
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.