Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551021
Description
NDUFA4 Polyclonal Antibody detects endogenous levels of NDUFA4 protein.
Cytochrome c oxidase (COX, complex IV) is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. Required for complex IV maintenance. {ECO:0000269; PubMed:22902835}.
Specifications
NDUFA4 | |
Polyclonal | |
Unconjugated | |
NDUFA4 | |
CI-9k; CI-MLRQ; Complex I 9kDa subunit; complex I-MLRQ; Cytochrome c oxidase subunit NDUFA4; EC 1.6.5.3; EC 1.6.99.3; MLRQ; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4 (9 kDa, MLRQ); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4; NADH-ubiquinone oxidoreductase MLRQ subunit; NDUFA4; NDUFA4, mitochondrial complex associated | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17992, 4697, 681024 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O00483, Q62425 | |
NDUFA4 | |
Synthesized peptide derived from the C-terminal region of human NDUFA4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction