Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA4L2 Monoclonal Antibody (OTI2E7)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527331

Catalog No. PIMA527331


Only null left
Add to Cart

Description

Description

NDUFA4L2 Monoclonal Antibody for Western Blot, IHC (P)

NDUFA4L2 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NDUFA4L2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NRX3
NDUFA4L2
Full length human recombinant protein of NDUFA4L2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2E7
Unconjugated
NDUFA4L2
BC064011; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2; NADH:ubiquinone oxidoreductase MLRQ subunit homolog; NADH-ubiquinone oxidoreductase MLRQ subunit homolog; NDUFA4, mitochondrial complex associated like 2; NDUFA4L2; NUOMS
Mouse
Affinity Chromatography
RUO
56901
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.