Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA4L2 Mouse anti-Human, Clone: 1G1H10, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 660501IG150UL
Description
NDUFA4L2 is a protein coding gene.Specifications
| NDUFA4L2 | |
| Monoclonal | |
| 1.06 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NRX3 | |
| NDUFA4L2 | |
| NDUFA4L2 Fusion Protein Ag9233 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1G1H10 | |
| Unconjugated | |
| NDUFA4L2 | |
| NDUFA4L2, NUOMS | |
| Mouse | |
| Protein A | |
| RUO | |
| 56901 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction