Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA5 Antibody (OTI1E8), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272904V
Description
NDUFA5 Monoclonal specifically detects NDUFA5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NDUFA5 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NDUFA5 | |
Recombinant protein fragment corresponding to amino acids 3-116 of human NDUFA5 (NP_004991) produced in HEK293T cells. | |
0.1 mL | |
4698.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1E8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
B13,5 (13kD, B13), DKFZp781K1356, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa, UQOR13FLJ12147 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction