Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA5 Antibody (OTI1E8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272904H
Description
NDUFA5 Monoclonal specifically detects NDUFA5 in Human samples. It is validated for Western Blot.Specifications
| NDUFA5 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| NDUFA5 | |
| Recombinant protein fragment corresponding to amino acids 3-116 of human NDUFA5 (NP_004991) produced in HEK293T cells. | |
| 0.1 mL | |
| 4698.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1E8 | |
| Western Blot | |
| B13,5 (13kD, B13), DKFZp781K1356, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa, UQOR13FLJ12147 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction