Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA5 Monoclonal Antibody (OTI1E8), TrueMAB™, OriGene
SDP

Catalog No. 502540659
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0659 30 μL
50-167-4664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0659 Supplier OriGene Supplier No. TA503881S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFA5 Monoclonal antibody specifically detects NDUFA5 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen NDUFA5
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1E8
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LPL
Gene Accession No. P06858
Gene Alias 2900002J19Rik; B13; CI-13kB; CI-13kD-B; complex I 13kDa subunit B; complex I subunit B13; Complex I-13kD-B; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH ubiquinone oxidoreductase subunit B13; NADH:ubiquinone oxidoreductase subunit A5; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; NADHUO; NDUFA5; NUFM; type I dehydrogenase; ubiquinone reductase; UQOR13
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 28-475 of human LPL produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4023
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.