Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118104
This item is not returnable.
View return policy
Description
NDUFA5 Polyclonal Antibody for Western Blot, IHC (P), IP
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFA5 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q16718 | |
| NDUFA5 | |
| E. coli-derived Human NDUFA5 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| NDUFA5 | |
| 2900002J19Rik; B13; CI-13kB; CI-13kD-B; complex I 13kDa subunit B; complex I subunit B13; Complex I-13kD-B; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH ubiquinone oxidoreductase subunit B13; NADH:ubiquinone oxidoreductase subunit A5; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; NADHUO; NDUFA5; NUFM; type I dehydrogenase; ubiquinone reductase; UQOR13 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 4698 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction