Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560270 Supplier Invitrogen™ Supplier No. PA560270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AMVKAEPDVK KLEDQLQGGQ LEEVILQAEH ELNLARKMRE WKLWEPLVEE PPADQWKWPI Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 78%.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFA5
Gene Accession No. Q16718, Q63362, Q9CPP6
Gene Alias 2900002J19Rik; B13; CI-13kB; CI-13kD-B; complex I 13kDa subunit B; complex I subunit B13; Complex I-13kD-B; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH ubiquinone oxidoreductase subunit B13; NADH:ubiquinone oxidoreductase subunit A5; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; NADHUO; NDUFA5; NUFM; type I dehydrogenase; ubiquinone reductase; UQOR13
Gene Symbols NDUFA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFA5. Recombinant protein control fragment (Product #RP-100136).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25488, 4698, 68202
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.