Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103944 Supplier Thermo Scientific Supplier No. PA5103944
Add to Cart
Add to Cart

NDUFA5 Polyclonal Antibody

NDUFA5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NDUFA5
Gene Accession No. Q16718, Q9CPP6
Gene Alias 2900002J19Rik; B13; CI-13kB; CI-13kD-B; complex I 13kDa subunit B; complex I subunit B13; Complex I-13kD-B; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH ubiquinone oxidoreductase subunit B13; NADH:ubiquinone oxidoreductase subunit A5; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; NADHUO; NDUFA5; NUFM; type I dehydrogenase; ubiquinone reductase; UQOR13
Gene Symbols NDUFA5
Host Species Rabbit
Immunogen A synthesized peptide derived from human NDUFA5(Accession Q16718), corresponding to amino acid residues P39-N89.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4698, 68202
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.