Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFA5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522143
Description
Recommended positive controls: H1299, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (82%), Rat (80%), Rhesus Monkey (94%), Chimpanzee (100%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
NDUFA5 | |
Polyclonal | |
Unconjugated | |
NDUFA5 | |
2900002J19Rik; B13; CI-13kB; CI-13kD-B; complex I 13kDa subunit B; complex I subunit B13; Complex I-13kD-B; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH ubiquinone oxidoreductase subunit B13; NADH:ubiquinone oxidoreductase subunit A5; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; NADHUO; NDUFA5; NUFM; type I dehydrogenase; ubiquinone reductase; UQOR13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4698, 68202 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M Tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q16718, Q9CPP6 | |
NDUFA5 | |
Full length human NDUFA5 Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction