Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570572 Supplier Invitrogen™ Supplier No. PA570572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 86%.

NDUFA6 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6), also known as LYRM6 (LYR motif-containing protein 6), NADHB14 or B14, is a 128 amino acid protein that localizes to the matrix side of the mitochondrial membrane and belongs to the complex I LYR family. Existing as a subunit of the multi-protein membrane respiratory chain NADH dehydrogenase complex (Complex I), NDUFA6 functions as an accessory protein that facilitates the transfer of electrons from NADH to the respiratory chain. The gene encoding NDUFA6 maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NDUFA6
Gene Accession No. P56556
Gene Alias 14kDa; 2700038D15Rik; B14; B230217P19Rik; CI-B14; complex I B14 subunit; complex I-B14; LOW QUALITY PROTEIN: NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 6; LYR motif-containing protein 6; LYRM6; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6 (B14); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6, 14kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 6; NADH:ubiquinone oxidoreductase subunit A6; NADHB14; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, 6; NADH-ubiquinone oxidoreductase B14 subunit; NDUFA6; zgc:56717
Gene Symbols NDUFA6
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NDUFA6 (aa 78-127).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4700
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.