Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA7 Antibody (OTI2G4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272909PCP
Description
NDUFA7 Monoclonal specifically detects NDUFA7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| NDUFA7 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| NDUFA7 | |
| Full length recombinant protein of human NDUFA7 (NP_004992) produced in HEK293T cells. | |
| 0.1 mL | |
| 4701.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2G4 | |
| Flow Cytometry | |
| B14.5a, CI-B14.5a, complex I B14.5a subunit, Complex I-B14.5a, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7 (14.5kD, B14.5a), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7, NADH-ubiquinone oxidoreductase subunit B14.5a | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction