Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA7 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
SDP

Catalog No. 502541361
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1361 30 μL
50-167-3737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1361 Supplier OriGene Supplier No. TA502701S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFA7 Monoclonal antibody specifically detects NDUFA7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen NDUFA7
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2E4
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CCNB1IP1
Gene Accession No. Q9NPC3
Gene Alias 14.5kDa; 2400007M02Rik; B14.5a; CI-B14.5a; complex I B14.5a subunit; complex I-B14.5a; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7 (B14.5a); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7; NADH:ubiquinone oxidoreductase subunit A7; NADH-ubiquinone oxidoreductase subunit B14.5a; Ndufa7
Host Species Mouse
Immunogen Full length human recombinant protein of human CCNB1IP1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57820
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.