Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFA7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563693
Description
Immunogen sequence: SATRLIQRLR NWASGHDLQG KLQLRYQEIS KRTQPPPKLP VGPSHKLSNN YYCTRDG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
NDUFA7 | |
Polyclonal | |
Unconjugated | |
Ndufa7 | |
14.5kDa; 2400007M02Rik; B14.5a; CI-B14.5a; complex I B14.5a subunit; complex I-B14.5a; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7 (B14.5a); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7; NADH:ubiquinone oxidoreductase subunit A7; NADH-ubiquinone oxidoreductase subunit B14.5a; Ndufa7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4701 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95182 | |
Ndufa7 | |
Recombinant protein corresponding to Human NDUFA7. Recombinant protein control fragment (Product #RP-99150). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction