Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536491 Supplier Invitrogen™ Supplier No. PA536491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 22 kDa. Purity is >95% by SDS-PAGE.

NDUFA8 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed to be not involved in catalysis. Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFA8
Gene Accession No. P51970, Q9DCJ5
Gene Alias 0610033L03Rik; Aa2-258; AW261656; CI-19KD; CI-PGIV; Complex I-19kD; complex I-PGIV; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8; NADH:ubiquinone oxidoreductase PGIV subunit; NADH:ubiquinone oxidoreductase subunit A8; NADH-ubiquinone oxidoreductase 19 kDa subunit; NDUFA8; PGIV
Gene Symbols NDUFA8
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 120-170 of human NDUFA8.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296658, 4702, 68375
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.