Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567275 Supplier Invitrogen™ Supplier No. PA567275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQLSKEAGVEK FIHVSHLNAN IKSSSRYLRN KAVGEKVVRD AFPEAIIVKP SDIFGREDRF LNSFASMHRF GPIPL Highest antigen sequence identity to the following orthologs - mouse 74%, rat 70%.

NDUFA9 (NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial) is also named as NDUFS2L, CI-39kD and belongs to the complex I NDUFA9 subunit family. It is involved in stabilizing the junction between membrane and matrix arms of complex I, a late assembly step critical for complex I biogenesis and activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFA9
Gene Accession No. Q16795
Gene Alias 1010001N11Rik; CC6; CI39k; CI-39k; CI-39kD; complex I 39kDa subunit; complex I-39kD; EMBL:AAH91192.1, ECO:0000312; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) 42 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial; NADH:ubiquinone oxidoreductase subunit A9; NADH-ubiquinone oxidoreductase 39 kDa subunit; NDUFA9; ndufa9 {ECO:0000312; NDUFS2L; RGD:1307307}; SDR22E1; short chain dehydrogenase/reductase family 22E, member 1; sperm flagella protein 3
Gene Symbols NDUFA9
Host Species Rabbit
Immunogen Recombinant Human NDUFA9. Recombinant protein control fragment (Product #RP-105577).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4704
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.