Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120861 Supplier Invitrogen™ Supplier No. PA5120861
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: LO2, BT-474, HL-60, HT-29, Mouse kidney, Mouse heart, Mouse brain, Mouse liver, Rat testis. The target is usually found in the following locations: Mitochondrion matrix. Immunogen sequence: MAAAAQSRVV RVLSMSRSAI TAIATSVCHG PPCRQLHHAL MPHGKGGRSS VSGIVATVFG ATGFLGRYVV NHLGRMGSQV IIPYRCDKYD IMHLRPMGDL GQLLFLEWDA RDKDSIRRVV QHSNVVINLI GRDWETKNFD FEDVFVKIPQ AIAQLSKEAG VEKFIHVSHL NANIKSSSRY LRNKAVGEKV VRDAFPEAII VKPSDIFGRE DRFLNSFASM HRFGPIPLGS LGWKTVKQPV YVVDVSKGIV NAVKDPDANG KSFAFVGPSR.

NDUFA9 (NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial) is also named as NDUFS2L, CI-39kD and belongs to the complex I NDUFA9 subunit family. It is involved in stabilizing the junction between membrane and matrix arms of complex I, a late assembly step critical for complex I biogenesis and activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFA9
Gene Accession No. Q16795, Q5BK63, Q9DC69
Gene Alias 1010001N11Rik; CC6; CI39k; CI-39k; CI-39kD; complex I 39kDa subunit; complex I-39kD; EMBL:AAH91192.1, ECO:0000312; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) 42 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial; NADH:ubiquinone oxidoreductase subunit A9; NADH-ubiquinone oxidoreductase 39 kDa subunit; NDUFA9; ndufa9 {ECO:0000312; NDUFS2L; RGD:1307307}; SDR22E1; short chain dehydrogenase/reductase family 22E, member 1; sperm flagella protein 3
Gene Symbols NDUFA9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human NDUFA9 (NP_004993.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362440, 4704, 66108
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.