Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988446
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8446 Supplier Bioss Supplier No. BS8301R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDUFA9 (NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial) is also named as NDUFS2L, CI-39kD and belongs to the complex I NDUFA9 subunit family. It is involved in stabilizing the junction between membrane and matrix arms of complex I, a late assembly step critical for complex I biogenesis and activity.

Specifications

Antigen NDUFA9
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NDUFA9
Gene Accession No. Q16795
Gene Alias 1010001N11Rik; BOS_6009; CC6; CI39k; CI-39k; CI-39kD; complex I 39kDa subunit; Complex I-39kD; EMBL:AAH91192.1, ECO:0000312; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) 42 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial; NADH:ubiquinone oxidoreductase subunit A9; NADH-ubiquinone oxidoreductase 39 kDa subunit; Ndufa9; ndufa9 {ECO:0000312; NDUFS2L; RGD:1307307}; SDR22E1; short chain dehydrogenase/reductase family 22E, member 1; Sperm flagella protein 3
Gene Symbols NDUFA9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NDUFA9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4704
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.