Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFAB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589967 Supplier Invitrogen™ Supplier No. PA589967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PALVLAQVPG RVTQLCRQYS DMPPLTLEGI QDRVLYVLKL YDKIDPEKLS VNSHFMKDLG LDSLDQVEII MAMEDEFGFE IPDIDAEKLM CPQEIVDYIA DKKDVYE Positive Samples: 293T, HT-29, U-87MG, LO2; Cellular Location: Mitochondrion.

Carrier of the growing fatty acid chain in fatty acid biosynthesis in mitochondria. Accessory and non-catalytic subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), which functions in the transfer of electrons from NADH to the respiratory chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFAB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene NDUFAB1
Gene Accession No. O14561, Q9CR21
Gene Alias 2210401F17Rik; 2310039H15Rik; 2610003B19Rik; 8kDa; 9130423F15Rik; ACP; Acyl carrier protein, mitochondrial; CI-SDAP; complex I SDAP subunit; FASN2A; mitochondrial acyl carrier protein; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa; NADH:ubiquinone oxidoreductase SDAP subunit; NADH:ubiquinone oxidoreductase subunit AB1; NADH-ubiquinone oxidoreductase 9.6 kDa subunit; NDUFAB1; SDAP
Gene Symbols NDUFAB1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293453, 4706, 70316
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.