Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFAB1 Recombinant Rabbit Monoclonal Antibody (JE65-76)
GREENER_CHOICE

Catalog No. PIMA544832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544832 Supplier Invitrogen™ Supplier No. MA544832
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NDUFAB1 Recombinant Monoclonal Antibody for Flow, IHC (P), Western Blot

Carrier of the growing fatty acid chain in fatty acid biosynthesis in mitochondria. Accessory and non-catalytic subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), which functions in the transfer of electrons from NADH to the respiratory chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFAB1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE65-76
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NDUFAB1
Gene Accession No. O14561
Gene Alias 2210401F17Rik; 2310039H15Rik; 2610003B19Rik; 8kDa; 9130423F15Rik; ACP; Acyl carrier protein, mitochondrial; CI-SDAP; complex I SDAP subunit; FASN2A; mitochondrial acyl carrier protein; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa; NADH:ubiquinone oxidoreductase SDAP subunit; NADH:ubiquinone oxidoreductase subunit AB1; NADH-ubiquinone oxidoreductase 9.6 kDa subunit; NDUFAB1; SDAP
Gene Symbols NDUFAB1
Host Species Rabbit
Immunogen Recombinant protein within human NDUFAB1 aa 57-156/156.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4706
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.